Would you like to log in again?
8270 Calibration Mix #2 (11 components), Restek
Supplier: Restek
31619
10118-476EA 70.03 USD InStock
10118-476
8270 Calibration Mix #2 (11 components), Restek
Standards Environmental Analysis Standards
Contains: Aniline (62-53-3); Benzidine (92-87-5); 4-Chloroaniline (106-47-8); 3,3'-Dichlorobenzidine (91-94-1); Diphenylamine (122-39-4)*; 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6); N-Nitrosodimethylamine (62-75-9); N-Nitrosodi-n-propylamine (621-64-7); Pyridine (110-86-1)
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Caution: *N-nitrosodiphenylamine (8270-listed analyte) decomposes to diphenylamine (mix component) in the injector.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty