Would you like to log in again?
Frosted Microscope Slides, Globe Scientific
Supplier: Globe Scientific
1324 1328 1304 1338 1334 1308
10118-592CS 653.88 USD InStock
10118-592 89500-472 89500-470 10118-596 10118-598 89500-474
Frosted Microscope Slides, Globe Scientific
Slides Microscope Slides
Globe Scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
  • Dimensions: 25×75×1 mm
  • Thickness: 1.1 mm
These frosted microscope slides are produced using a unique chemical etching process. This results in slides with a smooth frosted finish that won't wear out slide marking pens. Choose from single-sided or double-sided frostings, and ground or beveled edges. Ground-edge slides are also available with 90° corners (straight-edge), or safety corners (slightly clipped for comfort). All slides are pre-cleaned and supplied ready for use.
Packaging: Slides are packaged 72 slides per box, 20 boxes per case (10 gross).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty