Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DiamondⓇ White Glass Microscope Slides, Plain, Globe Scientific

Supplier: Globe Scientific
Diamond™
1380-10 1384-10
10118-618CS 901.62 USD InStock
10118-618 10118-612
DiamondⓇ White Glass Microscope Slides, Plain, Globe Scientific
Slides Microscope Slides
Globe scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.

  • Produced from the finest quality optical white glass
  • Dimensions: 25×75 mm
  • Thickness: 1 mm

DiamondⓇ White Glass plain microscope slides are available with 90° or clipped corners. The slides are pre-cleaned and supplied ready for use.

Packaging: Slides are packaged 72 per box, 20 boxes per case (10 gross).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty