Would you like to log in again?
Cover Glasses, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1405-10 1404-20 1408-20 1408-10 1418-10 1415-20 1412-10 1402-20 1418-20 1414-10 1415-15 1401-10 1419-10 1414-20 1411-20 1402-10 1411-10 1404-10 1407-20 1405-20 1404-15 1407-10 1415-10 1401-20 1412-20 1419-20
10118-639BX 71.6 USD InStock
10118-639 10118-655 10118-677 10118-675 10118-653 10027-469 10118-637 10118-657 10118-658 10118-663 10118-641 10118-661 10118-787 10118-645 10118-667 10118-642 10118-665 10118-649 10118-789 10118-647 10118-669 10118-791 10118-673 10118-651 10118-671 10118-793
Cover Glasses, Globe Scientific
Cover Glasses
Globe Scientific Cover Glasses are packaged 1 ounce per pack, 10 packs/box (10 oz.) Each flip-top pack contains a desiccant and is wrapped in aluminum to prevent sticking.
Cover Glasses are available in three thicknesses: No 1 (0.14mm), No 1.5 (0.175mm), and No 2 (0.21mm).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty