Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioBox, 81-Place, for 3, 4 and 5 ml CryoClear™ Vials, Globe Scientific

Supplier: Globe Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

3042B 3042Y 3042R 3042G
10118-738PK 102.93 USD InStock
10118-738 10118-740 10118-742 10118-736
BioBox, 81-Place, for 3, 4 and 5 ml CryoClear™ Vials, Globe Scientific
Boxes Cryoboxes
Produced from polycarbonate (PC), these storage boxes are designed to withstand temperatures of −196 to +121°C

  • Polycarbonate (PC)
  • Transparent lid for convenient viewing of contents
  • The lid is printed with numbers for easy sample identification
  • Lid has two angled corners to ensure the correct alignment of the well numbers
  • Autoclavable at +121 °C, 15 psi (1 bar) for 20 minutes.
  • Fits in stainless steel storage racks

The lid is transparent for viewing contents and has a printed numerical grid for easy sample identification. Each box features two angled corners to guarantee proper alignment of the grid. A frosted writing area provides the perfect surface for smear-resistant marking.

The boxes fit in stainless steel freezer racks and have vent holes for drainage and rapid freezing

Ordering information: Holds 81 vials (9×9 format). Includes a free vial picker.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty