Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Charged Microscope Slides, White Glass, Globe Scientific

Supplier: Globe Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

1354Y 1358T 1358Y 1358N 1358P 1354N 1358D 1358W 1358G 1358L 1358A 1358B 1354W
89500-508CS 2238.75 USD InStock
89500-508 89500-518 89500-506 89500-516 89500-504 10118-832 10118-319 89500-502 89500-512 89500-500 89500-510 89500-498 89500-496
Charged Microscope Slides, White Glass, Globe Scientific
Slides Adhesive Slides
Charged slides provide an ideal surface for cell and tissue adhesion.

  • Produced from the finest quality optical white glass
  • Charged surface allows cells and tissues to adhere to the slide
  • Packed in high-quality white plastic boxes
  • Dimensions: 25×75 mm
  • Thickness: 1 to 1.2 mm

Diamond™ White Glass Microscope slides are made from chemically pure white glass sheets. The glass offers a low iron content which results in superior transparency, very low self-flourescence, and no color distortion. The special composition of the glass provides minimal oxidation, which ensures sample integrity during long-term storage. Increased hardness and molecular stability of the slide surface guarantees more accurate experimental results. All Diamond™ White Glass microscope slides are packaged in high quality plastic boxes and double cellophane wrapped to protect against moisture and foreign particles.

Charged Microscope Slides with Beveled Edges and 45° Clipped Corners are available in bright colors and are pre-cleaned and supplied ready for use. These slides are packed in high quality white plastic boxes.

These popular slides are being utilized in histology, cytology, and pathology departments where the convenience of an adhesive slide saves the user time and money.

Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty