Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CART HYDRAULIC LIQUID GAS CYLINDER
Supplier: WESCO INDUSTRIAL PRODUCTS, LLC
Wesco Hydraulic Liquid Cylinder Cart^ Comes with hand brake for use when transporting cylinders, (2) 10in polyurethane front wheels, (1) 8in polyurethane swivel steering wheel, Handles cylinders up to 20in in diameter and 70in in height, Capacity: 750 lbs. 71x54x66.25 inch
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CART HYDRAULIC LIQUID GAS CYLINDER 10118-860 240250 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty