Would you like to log in again?
SEMI-AUTO DUCT APPARTUS 115V
Supplier: KOEHLER INSTRUMENT COM INC
Semi-Automatic Constant Temperature Ductility Testing Machine^ Conforms to the specifications of: ASTM D113, D5892, D6084, Capacity: 3 molds with samples, Maximum Travel Length: 150 cm, Standard Traction Speed: 5 cm/min, Timer: 1-999 min, 219.1 x 48.3 x 40.6cm, 15V 60Hz.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEMI-AUTO DUCT APPARTUS 115V 10118-906 K80060 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty