Would you like to log in again?
Bovine Blood Agar, 5%
Supplier: Hardy Diagnostics
A188 A189 A19BX
10118-938PK 46.01 USD InStock
10118-938 75878-358 10153-948
Bovine Blood Agar, 5%
Media for Microbiology Dehydrated Media for Microbiology
For detection of organisms that may cause mastitis in dairy cattle.
(TSA with 5% Triple Washed Cow Blood) For detection of organisms that may cause mastitis in dairy cattle, such as Staphylococcus aureus and Streptococcus agalactiae.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty