Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant Cyclophilin A (from E. coli)

Supplier: BioVendor
RD172329100
101189-698EA 415.82 USD InStock
101189-698
Human Recombinant Cyclophilin A (from E. coli)
Proteins and Peptides
Cyclophilins belong to a group of proteins that have peptidyl-prolyl cis-trans isomerase activity; such proteins are collectively known as immunophilins and also include the FK-506-binding proteins and the parvulins. Cyclophilins are found in all cells of all organisms studied, in both prokaryotes and eukaryotes. Human have a total of 16 cyclophilin proteins. Cyclophilins also have varying degrees of affinity for the immunosuppressive drug Cyclosporine A (CsA), a cyclic 11-amino-acid peptide produced by fungus Tolypocladium infantum. Cyclophilin A, in particular, is the major intracellular receptor for CsA. Cyclophilin A (CYPA) is the first member of the cyclophilins to be identified in mammals. Human genes of CYPA, also known as Cyp18, are located on chromosome 7p11.2-p13 and encode the protein, which consists of 165 amino acid residues with a relative molecular mass approximately 18 kDa. Human CYPA has an eight-stranded antiparallel β-barrel structure, with two α-helices enclosing the barrel from either side. Seven aromatic and other hydrophobic residues form a compact hydrophobic core within the barrel, usually in the area where CsA binds. A loop from Lys118 ti His126 and four β-strands (β3-β6) make up the binding site for CsA. In mammals, the CsA-CYPA complex binds to and inhibits calcineurin, a calcium-calmodulin-activated serine/threonine-specific protein phosphatase. The inhibition of calcineurin blocks the translocation of nuclear factor of activated T cells from the cytosol to the nucleus, thus preventing the transcription of genes encoding cytokines such as interleukin-2.

Sequence: MKHHHHHHAS VNPTVFFDIA VDGEPLGRVS FELFADKVPK TAENFRALST GEKGFGYKGS CFHRIIPGFM CQGGDFTRHN GTGGKSIYGE KFEDENFILK HTGPGILSMA NAGPNTNGSQ FFICTAKTEW LDGKHVVFGK VKEGMNIVEA MERFGSRNGK TSKKITIADC GQLE

Other names: Peptidyl-prolyl cis-trans isomerase A, PPIase A, Cyclosporin A-binding protein, Rotamase A, PPIA, CYPA

Ordering information: For research use only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty