Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Pyruvate Kinase M2 (from E. coli)

Supplier: BioVendor
RD172345100
101189-702EA 415.82 USD InStock
101189-702
Human Pyruvate Kinase M2 (from E. coli)
Enzymes
Pyruvate kinase is a glycolytic enzyme

Sequence: MKHHHHHHAS KP­HSEAGTAF IQTQQLHAAM ADTFLEHMCR LDIDSPPITA RNTGIICTIG PASRSVETLK EMIKSGMNVA RLNFSHGTHE YHAETIKNVR TATESFASDP ILYRPVAVAL DTKGPEIRTG LIKGSGTAEV ELKKGATLKI TLDNAYMEKC DENILWLDYK NICKVVEVGS KIYVDDGLIS LQVKQKGADF LVTEVENGGS LGSKKGVNLP GAAVDLPAVS EKDIQDLKFG VEQDVDMVFA SFIRKASDVH EVRKVLGEKG KNIKIISKIE NHEGVRRFDE ILEASDGIMV ARGDLGIEIP AEKVFLAQKM MIGRCNRAGK PVICATQMLE SMIKKPRPTR AEGSDVANAV LDGADCIMLS GETAKGDYPL EAVRMQHLIA REAEAAIYHL QLFEELRRLA PITSDPTEAT AVGAVEASFK CCSGAIIVLT KSGRSAHQVA RYRPRAPIIA VTRNPQTARQ AHLYRGIFPV LCKDPVQEAW AEDVDLRVNF AMNVGKARGF FKKGDVVIVL TGWRPGSGFT NTMRVVPVP

Other names: Pyruvate kinase isozymes M1/M2, Cytosolic thyroid hormone-binding protein, CTHBP, Opa-interacting protein 3, OIP-3, Pyruvate kinase 2/3, Pyruvate kinase muscle isozyme, Thyroid hormone-binding protein 1, THBP1, Tumor M2-PK, PKM, OIP3, PK2, PK3, PKM2

It catalyzes the transfer of a phosphoryl group from phosphoenolpyruvate to ADP, which results in ATP formation. Isoenzyme M2 is expressed in early fetal tissues and in cancer cells.

Ordering information: For research use only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty