Would you like to log in again?
Human Hiv-1 Protease (from E. coli)
Supplier: BioVendor
RH1P0001
101189-714EA 776.77 USD InStock
101189-714
Human Hiv-1 Protease (from E. coli)
Enzymes
Retroviral protease from HIV-1 virus is an enzyme important in the life cycle of the virus
Sequence: PQITLWQRPL VTIKIGGQLK EALLDTGADD TVLEEMNLPG RWKPKMIGGI GGFIKVRQYD QILIEICGHK AIGTVLVGPT PVNIIGRNLL TQIGCTLNF
It is expressed in the infected cells as a part of Gag-Pol polyprotein from which it is autocatalytycaly released after formation of immature viral particle. The enzyme subsequently cleaves the other parts of viral polyproteins causing the maturation of the virus. In HIV-infected patients the enzyme is a subject of intensive mutagenesis and mutants resistant to applied medcines are produced as a consequence of the seletion pressure. HIV-1 protease is active as a homodimer.
Ordering information: For research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty