Would you like to log in again?
Anti-KLH Rat IgG ELISA, BioVendor
Supplier: BioVendor
636-13759
101189-752EA 1220.85 USD InStock
101189-752
Anti-KLH Rat IgG ELISA, BioVendor
Assays ELISAs
Keyhole limpet hemocyanin (KLH) is a large cooper-containing protein consisting of subunits with MW of 400 kDa.
  • Calibration Range: 0.47–30 ng/mL
  • Limit of Detection: 47 pg/mL
  • Plasma, Serum: 50 uL/well
Keyhole limpet hemocyanin (KLH) is a large cooper-containing protein consisting of subunits with MW of 400 kDa. It is found in the hemolymph of the sea mollusk Megathura crenulata. This extracellular respiratory protein has many immunostimulatory properties, including the ability to enhance the host’s immune response by interacting with T cells, monocytes, macrophages, andpolymorphonuclear lymphocytes. Since its discovery, KLH has been used primarily as a carrier for vaccines and antigens and as adjuvant treatment in regimens such as antimicrobial therapy.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty