Would you like to log in again?
Human Peripheral Blood Mononuclear Cells (PBMC), Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PB009C-50 PB009C-3 PB009C-2 PB009C-1
101189-982EA 248.08 USD InStock
101189-982 101189-980 101189-984 102830-054
Human Peripheral Blood Mononuclear Cells (PBMC), Charles River Laboratories Cell Solutions
Blood
Human peripheral blood mononuclear cells (PBMC) are collected from healthy human volunteer donors following cGMP and cGTP collection guidelines under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol
Mononuclear cells are widely used in research and clinical applications such as in microbiology, virology, oncology, vaccine development, transplant and regenerative biology, and toxicology.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty