Would you like to log in again?
Human Peripheral Blood CD3 Pan T Cells, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB03NC-1 PB03NC-2 PB03NC-3 PB03C-3 PB03C-2 PB03C-1
102830-058EA 1362.4 USD InStock
102830-058 102830-060 101190-000 101190-002 102830-056 101189-998
Human Peripheral Blood CD3 Pan T Cells, Charles River Laboratories Cell Solutions
Blood
Human peripheral blood CD3 Pan T Cells are isolated from peripheral blood mononuclear cells (PBMCs) using either POSITIVE or NEGATIVE immunomagnetic cell separation procedures.
PBMCs are collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty