Would you like to log in again?
Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5643 5641 56431
10119-366CS 1788.99 USD InStock
10119-366 89404-878 10119-368
Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads, Thermo Scientific
Spill Control Materials
Absorb and lock in liquids with Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads.
  • Fiber resists tearing, even when saturated
  • Ideal for autopsy work
  • Reduced exposure
Absorb and lock in liquids with Thermo Scientific™, Shandon™, Suresorb™, Clinical Pads and Formaldehyde Neutralizer Pads. Ideal for containing and disposing of body fluids containing Hepatitis B virus, AIDS / HIV and other pathogens. Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors, ideal for grossing areas and autopsy work. Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty