Would you like to log in again?
MultiWash™+ Microplate Washer, Molecular Devices
Supplier: Molecular Devices
MULTIWASH+ MULTIWASH+
10119-372EA 13553.49 USD InStock
10119-372 CA10119-372
MultiWash™+ Microplate Washer, Molecular Devices
Microplate Washers
A compact, quiet, automated washer with walk-up usability.
  • User-friendly
  • Include 20 preset protocols
  • Comes with 4 wash bottles
The user-friendly MultiWash™+ Microplate Washer from Molecular Devices, one of the pioneers in microplate detection, includes 20 preset protocols, works with both 96- and 384-well plates, and comes with 4 different wash/rinse bottles for out-of-the-box use
MultiWash™+ Microplate Washer benefits include, walk-up usability, 4 wash and rinse bottles included, the flexibility to wash 96- or 384-well plates, automated rinse to reduce clogging, and cross-wise aspiration to reduce residual volume within each well. The device is vacuum- and pressure-free. On-board pumps provide a quiet wash experience.
The MultiWash™+ Microplate Washer features a variety of wash protocols and modes. An on-board touch panel offers the option of 20 different wash protocols, with up to 8 cycles within each protocol. The unit also provides adjustable speed and volume, adjustable aspiration speed and time, and adjustable soak times. Three shaking modes are available.
The unit is simply a great fit for any lab.
Ordering information: The MultiWash™+ Microplate Washer includes the MultiWash™+ for 96-well plates, 4 liquid lines, 8-way manifold and tubing, 4 wash/rinse bottles (2L), and 1 waste bottle (2.5L). Additional options include, optional 16-way manifold for 384-well plates, optional 12-way manifold, optional washer maintenance kit (2XT 2.5A spare fuses, 2 cleaning needles, tubing sets for all bottles, cleaning tool for needles, tubing set for manifold, and spare tubing set for manifold), optional external tubing kit (4-way), and optional replacement wash/rinse bottles (2L). Contact your Avantor sales representative for additional information on all optional items.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty