Would you like to log in again?
AnaeroGRO™ Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
AG901 AG313 AG601 AG401 AG701 AG22H AG23H AG303 AG061 AG304 AG303 AG301 AG312 AG302
10145-976PK 17.08 USD InStock
10145-976 10324-350 10324-360 10127-480 10324-354 10324-364 10324-352 10324-362 10324-358 10324-346 10324-356 10324-366 10119-762 CA10001-000
AnaeroGRO™ Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and differentiation of anaerobic bacteria.
  • Enriched, selective, and differential media
  • Cultivate anaerobic bacteria
  • Selective isolation of gram-positive and gram-negative bacteria
Hardy Diagnostics AnaeroGRO™ Anaerobic PEA (Phenylethyl Alcohol) Agar is recommended for use as an enriched and selective medium for the cultivation and selective isolation of gram-positive and negative obligate anaerobic bacteria
It is useful in isolating obligate anaerobes from mixed flora, by inhibiting gram-negative facultative anaerobes and the control of swarming organisms. Hardy Diagnostics AnaeroGRO™ Bacteroides Bile Esculin Agar is an enriched, selective, and differential medium recommended for the rapid isolation and presumptive identification of obligately anaerobic gram-negative bacilli of the Bacteroides fragilis group and Bilophila spp.
Hardy Diagnostics AnaeroGRO™ LKV (Laked Blood with Kanamycin and Vancomycin) Agar is recommended for use in the selective isolation and partial identification of obligately anaerobic gram-negative bacilli, such as Prevotella spp. and Bacteroides spp.
Hardy Diagnostics AnaeroGro™ Brucella Agar with Hemin and Vitamin K is recommended for use in the primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens. The medium is also suitable for the growth of aerobic and microaerophilic bacteria when incubated under the appropriate conditions.
Hardy Diagnostics AnaeroGRO™ Thioglycollate with Hemin and Vitamin K (H and K) is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty