Would you like to log in again?
Human Mobilized Peripheral Blood CD133 Stem / Progenitor Cells, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M133C-2 M133C-3 M133C-1
101222-934EA 7223.67 USD InStock
101222-934 101222-936 101190-074
Human Mobilized Peripheral Blood CD133 Stem / Progenitor Cells, Charles River Laboratories Cell Solutions
Blood
Human mobilized peripheral blood CD133 stem/progenitor cells are isolated from peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic cell separation procedures.
PBMCs are collected from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. Donors are injected with G-CSF (Neupogen® - FDA approved), which stimulates the bone marrow to produce a large number of hematopoietic and progenitor stem cells and mobilizes them into the peripheral blood stream. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty