Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CritiCore ESD Stripe Cleanroom Hoods with Open Face Under Chin
Supplier: CritiCore Protective Wear
CritiCore
10120-388 10120-404 10120-386 10120-394 10120-384 10120-408 10120-378 10120-398 10120-382 10120-392 10120-406 10120-380 10120-402 10120-400 10120-396 10120-390
10120-380EA 24.28 USD InStock
10120-380 10120-392 10120-390 10120-408 10120-406 10120-404 10120-402 10120-388 10120-378 10120-400 10120-386 10120-398 10120-384 10120-396 10120-382 10120-394
CritiCore ESD Stripe Cleanroom Hoods with Open Face Under Chin
Workwear
CritiCore’s reusable, controlled environment ESD stripe hoods are suitable for a variety of cleanroom applications, including pharmaceutical, biotechnology, and medical device manufacturing.
  • Open face design under the chin
  • Made with a high-density polyester calendered fabric that resists fluids and bacterial penetration
  • Hoods feature a tunneled elastic back plus horizontal and vertical adjustment snaps
  • Split-drape design with a front panel that is longer than the back
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
  • Available in white and navy
  • Sizes range from XS to 4XL
Designed for cleanroom applications up to and including Class 10 (ISO 4), these hoods also include a one percent carbon stripe for electrostatic dissipation.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty