Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid 0.01 Acetous titrant

Supplier: 0000019890

6603-1L
101203-014EA 87.35 USD InStock
101203-014
Perchloric acid 0.01 Acetous titrant
Perchloric acid


Formula: HClO₄
MW: 100.46 g/mol
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1760
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty