Would you like to log in again?
Human Cystatin C ELISA, Biovendor
Supplier: BioVendor
RD191009100
101215-160EA 632.77 USD InStock
101215-160
Human Cystatin C ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of cystatin C.
In the BioVendor Human Cystatin C ELISA, standards, quality controls and samples are incubated in microtitrate plate wells pre-coated with polyclonal anti-human cystatin C antibody. After 30 min incubation and washing, polyclonal anti-human cystatin C antibody, conjugated with horseradish peroxidase (HRP) is added to the wells and incubated for 30 min with captured cystatin C. Following another washing step, the remaining HRP conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of cystatin C. A standard curve is constructed by plotting absorbance values against concentrations of cystatin C standards, and concentrations of unknown samples are determined using this standard curve.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty