Would you like to log in again?
PMN Elastase Human ELISA, BioVendor
Supplier: BioVendor
RM191021100
101215-164EA 787.77 USD InStock
101215-164
PMN Elastase Human ELISA, BioVendor
Assays ELISAs
PMN elastase is localised in the azurophilic granules of the polymorphonuclear granulocytes.
  • Calibration Range: 15.6-1000 ng/mL
  • Limit of Detection: 0.2 ng/mL
  • Bronchoalveolar lavage, Cerebrospinal fluid, Exudate, Plasma-Citrate, Plasma-EDTA, Seminal plasma: 10 uL/well
The human organism reacts with an inflammatory response to attacks of invading pathogens (micro-organisms and viruses) or damaged tissue (after accidents or surgery). Polymorphonuclear (PMN) granulocytes play an important role as primary defence cells in this inflammatory reaction. Different bloodstream mediators (cytokines, leukotrienes, complement factors, bacterial endotoxins, clotting and fibrinolysis factors) attract and stimulate these cells to phagocytize and destroy not naturally occurring agents. PMN granulocytes use proteinases to digest these agents and tissue debris. One of these proteinases is PMN elastase which is localised in the azurophilic granules of the polymorphonuclear granulocytes. During phagocytosis of foreign substances these enzymes are also partially excreted into the extracellular surrounding, where the aktivity of PMN elastase is regulated by inhibitors (esp. the α1-proteinase inhibitor, α1-PI). An overwhelming release of PMN elastase, however, can exceed the inhibitory potential of the α1-proteinase inhibitor. Thus, enzymatically active PMN elastase, together with simultaneously produced oxidants (O2-radicals, H2O2, OH-radicals), can cause local tissue injury. Due to the bloodstream and lymphatic system, however, α1-PI is delivered subsequently and eventually able to form a complex with all excreted elastase. Therefore, the concentration of the PMN elastase/ α1-PI complex correlates with the released PMN elastase and can be used as a measure for the activity of granulocytes during an inflammatory response. Primarily, determinations of PMN elastase find its application in observation of the course of trauma, shock and sepsis. Further indications are the areas of hemodialysis, infections by obstetrics, joint diseases, effusions of sport injuries, intestinal affection, pancreatitis, cystic fibrosis and male adnex affections.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty