Would you like to log in again?
Fetuin-A (AHSG) Human ELISA, Biovendor
Supplier: BioVendor
RD191037100
101215-170EA 816.92 USD InStock
101215-170
Fetuin-A (AHSG) Human ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of fetuin-A.
In the BioVendor Human Fetuin-A ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with polyclonal anti-human fetuin-A antibody. After 60 min incubation and washing, polyclonal anti-human fetuin-A antibody, conjugated with horseradish peroxidase (HRP) is added to the wells and incubated for 60 min with captured fetuin-A. Following another washing step, the remaining HRP conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of fetuin-A. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty