Would you like to log in again?
Leptin Receptor Human ELISA, BioVendor
Supplier: BioVendor
RD194002100
101215-182EA 816.92 USD InStock
101215-182
Leptin Receptor Human ELISA, BioVendor
Assays ELISAs
Leptin receptor (OB R) was identified as a leptin binding protein.
  • Calibration Range: 2-100 ng/mL
  • Limit of Detection: 0.04 ng/mL
  • Plasma-EDTA, Plasma-Heparin, Serum: 35 uL/well
Leptin receptor (OB R) was identified as a leptin binding protein (Leptin, the product of the ob gene, is a single chain 16 kDa protein consisting of 146 amino acid residues.) OB R was found to be a member of the class I cytokine receptor family with a large extracellular domain comprising 816 amino acid residues. Leptin receptor exists in multiple forms with a common extracellular domain and a variable length cytoplasmatic portion. Alternate splicing from a single gene derives the six isoforms of the Leptin receptor. The soluble form of the Leptin receptor, OB R contains no intracellular motifs or transmembrane residues, thus it consists entirely of the extracellular ligand binding domain of the receptor. Long forms of OB-R transcripts were reported to be expressed predominantly in regions of the hypothalamus which provides evidence that Leptin receptor is important in body weight regulation. Expression of short forms of OB-R transcripts have been found in multiple tissues, including the choroid plexus, lung, kidney, and primitive hematopoietic cell populations. Leptin receptor may act as a negative regulator of Leptin activity and it may maintain a pool of available bioactive Leptin by binding and delaying its clearance from circulation. Soluble Leptin receptor levels are indirectly proportional to adiposity and are increased in females versus males. Leptin receptor levels are highest in infants, decrease into adolescence, and remain relatively stable throughout adulthood. Soluble Leptin receptor is also found upregulated in patients with chronic heart failure, end-stage renal disease and anorexia.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty