Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Osteoprotegerin Human ELISA, BioVendor

Supplier: BioVendor
RD194003200
101215-184EA 919.38 USD InStock
101215-184
Osteoprotegerin Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of Osteoprotegerin.

  • Calibration Range: 2-100 ng/ml
  • Limit of Detection: 0.04 ng/ml
  • Plasma-EDTA, Plasma-Heparin, Serum: 35 ul/well

The RD194003200 Human Osteoprotegerin ELISA is a sandwich enzyme immunoassay for the quantitative measurement of human osteoprotegerin.

Osteoprotegerin (OPG, osteoclastogenesis inhibitory factor, OCIF) is a product of the TNFRSF11B gene, located on chromosome 8q24. OPG belongs to the TNF (tumor necrosis factor receptor) superfamily, that plays a key role in bone remodeling. Human OPG is a secreted glycoprotein composed of 401 aminoacid residues. OPG exists as a disulfide-linked homodimer (120 kDa) or as a monomer (60 kDa). Both of these forms are active but the dimer is more bioactive than the monomer. In contrast to most members of the TNF receptor superfamily, OPG probably exists only in a soluble form. Its ligands are RANKL and TRAIL. Human OPG shares 85% aminoacid identity to mouse OPG and 86 % identity to rat OPG. In adult humans OPG mRNA is highly expressed in bones (osteblasts), endothelial vessel cells, skin, liver, stomach, intestine, heart, brain and lung and is also present in atherosclerotic plaques.

In the BioVendor Human Osteoprotegerin ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with monoclonal anti-human OPG antibody. After 60 minutes incubation and washing, biotin labelled polyclonal anti-human OPG antibody is added and incubated for 60 minutes with captured OPG. After another washing, streptavidin-HRP conjugate is added. After 30 minutes incubation and the last washing step, the remaining conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of OPG. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Caution: This is intended for research use only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty