Would you like to log in again?
sTfR Human ELISA, (Soluble Transferrin Receptor), BioVendor
Supplier: BioVendor
RD194011100
101215-186EA 632.77 USD InStock
101215-186
sTfR Human ELISA, (Soluble Transferrin Receptor), BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of sTFr.
  • Calibration Range: 1.5 - 60 pmol/L
  • Limit of Detection: 0.03 pmol/L
  • Plasma-citrate, plasma-EDTA, plasma-heparin, serum: 35 µl/well
The RD194011100 Human sTfR ELISA is a sandwich enzyme immunoassay for the quantitative measurement of human soluble transferrin receptor. The transferrin receptor (TfR) is the gateway for transferrin-bound-iron entering all body cells. TfR is abundant on the surface of many newly formed cells, but the erythroid marrow cells account for 70 to 80 % of the total body TfR content. The soluble (or serum) transferrin receptor (sTfR) is a circulating truncated form of the membrane receptor protein; it is an 85 kDa glycoprotein forming in serum a 320 kDa complex with diferric transferrin. The serum sTfR concentration reflects the total body mass of cellular transferrin receptor. In the BioVendor Human sTfR ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with monoclonal anti-human sTfR antibody. After 60 minutes incubation and washing, monoclonal anti-human sTfR antibody, conjugated with horseradish peroxidase (HRP) is added to the wells and incubated for 60 minutes with captured sTfR. Following another washing step, the remaining HRP conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of sTfR. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty