Would you like to log in again?
Leptin Mouse/Rat ELISA, BioVendor
Supplier: BioVendor
RD291001200
101215-202EA 528.92 USD InStock
101215-202
Leptin Mouse/Rat ELISA, BioVendor
Assays ELISAs
Leptin is a protein hormone with important effects in metabolism and regulating body weight.
  • Calibration Range: 100-4000 pg/mL
  • Limit of Detection: 50 pg/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 7 uL/well
Leptin is a protein hormone with important effects in metabolism and regulating body weight. It is a single-chain 16 kDa protein consisting of 146 amino acid residues and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, small amounts of leptin are also secreted by cells in the epithelium of stomach and in the placenta. Leptin´s effect on body weight is mediated through effects on hypothalamic centers, where leptin receptors are highly expressed. Leptin has a dual action, it decreases the appetite and increases energy consumption.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty