Would you like to log in again?
Nexera X2 Series HPLC/UHPLC, Shimadzu
Supplier: Shimadzu
228-45185-58 228-45175-42 228-45180-58 228-45167-58 228-45166-58 228-45209-44 228-45180-57 228-45160-42 228-45178-42
10122-346EA 15458.73 USD InStock
10122-346 10122-344 10122-520 10122-428 10122-416 10122-426 10122-418 10122-342 10122-340
Nexera X2 Series HPLC/UHPLC, Shimadzu
Autosamplers
Shimadzu’s Nexera X2 Series delivers superior, real-world performance across a wider application range while delivering unparalleled flexibility and reliability in HPLC/UHPLC analyses
Key features include ultrahigh-throughput and clean autosampler–perfect front-end systems for all LCMS platforms, automated solvent blending and method scouting–ideal for method development, new PDA detector–offers superior sensitivity and resolution, achieving a 0.4 × 10-5 AU noise level for genuine UHPLC analysis, intelligent peak deconvolution analysis (i-PDeA) function–unique software solution to separate and accurately integrate co-eluting peaks and intelligent dynamic range extension calculator (i-DReC), which extends the dynamic range so low concentration (trace or impurity compounds) and very high concentration samples can be accurately analyzed in a single injection.
The Nexera X2 is optimized to enable analysis at pressures up to 19,000 psi without compromising performance and data quality. Nexera X2 autosamplers provide near-zero carryover, support ultrafast analysis with the world’s fastest cycle time (14 secs) and deliver excellent injection reproducibility down to 0.1 μL injection volumes. In addition, the Nexera X2 systems can be configured with a variety of solvent delivery units and column ovens to meet any laboratory workflow.
Certifications: UL Listed and CSA Listed.
Ordering information: To order, contact your Avantor sales representative.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty