Would you like to log in again?
FCV-130AL LC-8A
Supplier: SHIMADZU SCIENTIFIC
FCV-130al, LC-8a^ FCV-130al, LC-8a, reservoir select valve operates under the control of the LC-8a pump and has standard access to two reservoirs with the option for a third reservoir
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FCV-130AL LC-8A 10122-482 228-45078-91 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty