Would you like to log in again?
STAINLESS ELECTRIC HI-LIFT 27X46 FORKS
Supplier: WESCO INDUSTRIAL PRODUCTS, LLC
Truck^ Wesco* stainless steel battery powered high-lift skid truck 27x46 2200 lb for corrosion-resistant and clean room, forks: 27inchw x 46inch, 12 volt battery, 6.5inch wide forks, 8inch raised height, 3inch lowered height, 329 lbs., nylon steering and load wheels
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STAINLESS ELECTRIC HI-LIFT 27X46 FORKS 10122-526 272860 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty