Would you like to log in again?
TruLine ORP 50 Electrode, YSI
Supplier: YSI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TruLine
400355
10122-562EA 373.6 USD InStock
10122-562
TruLine ORP 50 Electrode, YSI
Electrodes Redox Electrodes
TruLine ORP 50, ORP electrode, Single Junction, Refillable, Glass shaft, 1 m cable, BNC Connector.
Ag/AgCl Reference System. SMEK plug head.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty