Would you like to log in again?
TruLab 1320 Dual Channel Laboratory Instrument for pH, mV (ORP) and Temperature, YSI
Supplier: YSI
1GA340Y
10122-576EA 2774.3 USD InStock
10122-576
TruLab 1320 Dual Channel Laboratory Instrument for pH, mV (ORP) and Temperature, YSI
Electrochemical Meters pH and Multi-Parameter Meters
The YSI TruLab® line includes the 1110 (single channel), 1310 and 1310P (single channel) and 1320 and 1320P (dual channel) instruments providing easy-to-use and calibrate instruments ideal for the laboratory.
Measure pH, mV (ORP) and temperature accurately in the laboratory with the YSI pH 1320. This benchtop instrument for basic, routine measurements in the laboratory is an accurate and easy-to-use solution for repeatable pH or mV measurements.
Repeatable/Efficient–The instrument's AutoRead function indicates when a stable measurement is reached improving repeatability.
ISE measurements include Blank value correction, increment methods: Known addition (single); Known subtraction; Sample addition; Sample subtraction; Known addition (double).
Certifications: CE, cETLus, electrical protective class III, IP43.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty