Would you like to log in again?
TruLine pH 25 Electrode, YSI
Supplier: YSI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
400353
10122-588EA 220.46 USD InStock
10122-588
TruLine pH 25 Electrode, YSI
Electrodes pH Electrodes
TruLine pH 25, pH electrode, Single Junction, Combination, Gel electrolyte, Epoxy shaft, 1 m cable, BNC Connector.
BNC head.
Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty