Would you like to log in again?
Human Peripheral Blood Macrophages (Mixed), Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PBMACC-MON-3 PBMACC-MON-2 PBMACC-MON-1
101190-048EA 2823.93 USD InStock
101190-048 101190-050 101222-950
Human Peripheral Blood Macrophages (Mixed), Charles River Laboratories Cell Solutions
Blood
Human peripheral blood macrophages (mixed) is a 50%-50% mixture of M1 and M2 macrophages after harvest
PBMCs are collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
M1 and M2 macrophages are derived from purified monocytes. CD14 monocytes were isolated from peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic cell separation procedures. M1 macrophages are cultured in the presence of GM-CSF for 10 days and M2 macrophages are cultured in the presence of M-CSF for 10 days. After 10 days, macrophages are combined at a 50%-50% ratio.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty