Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.1 M traceable to NIST Standard Reference Material (SRM)

Supplier: AQUA SOLUTIONS MS
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

8625-20L 8625-1L 8625-10L 8625-4L 8625-500ML 8625-4LC
101223-406EA 17.88 USD InStock
101223-406 101223-190 101223-140 101225-214 101223-036 101223-014
Sodium hydroxide 0.1 M traceable to NIST Standard Reference Material (SRM)
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
Order Now
Specification Test Results
Normality 0.1000 ± 0.0005 Normal @ 20° C
Appearance and Color Clear Water White Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty