Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% ACS

Supplier: AQUA SOLUTIONS MS
Synonyms: Muriatic acidHydrochloric acid solution

H2505-500ML H2505-2.5L H2505-2.5L(PVC) H2505-4L
101224-404EA 109.26 USD InStock
101224-404 101223-072 101226-512 101223-970
Hydrochloric acid 37% ACS
Hydrochloric acid
Appearance Colorless fuming liquid
Harmonization Code 2806.10.0000


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 36.5 – 38.0%
Color (APHA) Max. 10
Residue after Ignition Max. 5 ppm
Bromide (Br) Max. 0.005%
Free chlorine (Cl) Max. 1 ppm
Ammonium (NH4) Max. 3 ppm
Arsenic (As) Max. 0.01 ppm
Heavy metals (as Pb) Max. 1 ppm
Iron (Fe) Max. 0.2 ppm
Sulfate (SO4) Max. 1 ppm
Sulfite (SO2) Max. 1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty