Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

n-Heptane, high purity

Supplier: 0000019890
Danger

H1903-4L H1903-20L H1903-1L H1903-500ML
101224-678EA 355.87 USD InStock
101224-678 101225-382 101225-444 101223-476
n-Heptane, high purity
n-Heptane


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now
Specification Test Results
Assay (as n-Heptane) 99.0% Min
Assay (as C7 Hydrocarbons) 99.5% Min.
Assay (Total Hydrocarbons) 99.9% Min
Specific Gravity @ 15.56C 0.680 - 0.695
Titrable Acid 0.0003 meq/g Max
Residue After Evaporation 10 ppm Max.
Sulfur Compounds (as S) 0.005% Max
Water 0.03% Max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty