Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.1 N in isopropanol (IPA)

Supplier: AQUA SOLUTIONS MS
Danger
Synonyms: Muriatic acid

4131-4L 4131-20L 4131-500ML 4131-1L 4131-250ML 4131-4LC
101225-198EA 496.38 USD InStock
101225-198 101223-060 101223-174 101226-788 103194-120 101225-234
Hydrochloric acid 0.1 N in isopropanol (IPA)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Actual Normality 0.0995 - 0.1005

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty