Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.1 N in aqueous solution

Supplier: AQUA SOLUTIONS MS
Danger
Synonyms: Muriatic acidHydrochloric acid solution

4130-1L 4130-4LC 4130-4L 4130-10L 4130-20L 4130-4LG 4130-500ML
102465-500EA 73.15 USD InStock
102465-500 101224-038 101226-922 101223-086 101226-446 101226-798 101222-990
Hydrochloric acid 0.1 N in aqueous solution
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty