Would you like to log in again?
Protein G Magnetic Beads, New England Biolabs
Supplier: New England Biolabs
S1430S S1430S
101227-250EA 234 USD InStock
CA101227-250 101227-250
Protein G Magnetic Beads, New England Biolabs
Beads Magnetic Beads
Protein G Magnetic Beads are an affinity matrix for the small-scale isolation and purification of immunoglobulins.
A truncated form of recombinant Protein G is covalently coupled to a nonporous paramagnetic particle. Protein G exhibits high affinity for subclasses of IgG from many species including human, rabbit, mouse, rat and sheep. In addition, Protein G can be used for immunoprecipitations with mouse monoclonal antibodies. The protein is coupled through a linkage that is stable and leak resistant over a wide pH range. This permits the immunomagnetic purification of IgGs from ascites, serum or cell culture supernatants; the matrix can then be regenerated without loss of binding capacity. Protein G Magnetic Beads can be used to immunoprecipitate target proteins from crude cell lysates using selected primary antibody. In addition, specific antibodies can be chemically cross-linked to the Protein G coated surface to create a reusable immunoprecipitation bead, avoiding the co-elution of antibody with target antigen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty