Would you like to log in again?
DEEP VENT™ (exo-) DNA Polymerase, New England Biolabs
Supplier: New England Biolabs
DEEP VENT™
M0259L M0259L M0259S M0259S
101227-262EA 120 USD InStock
CA101227-260 101227-262 CA101227-262 101227-260
DEEP VENT™ (exo-) DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Deep VentR (exo-) DNA Polymerase has been genetically engineered to eliminate the 3´→ 5´ proofreading exonuclease activity associated with Deep Vent DNA Polymerase
Deep Vent (exo-) DNA Polymerase is even more stable than VentR® (exo-) DNA Polymerase with a half-life of 23 hours at 95°C and 8 hours at 100°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty