Would you like to log in again?
Enterokinase
Supplier: New England Biolabs
P8070S P8070L P8070L P8070S
101227-278EA 138 USD InStock
CA101227-276 101227-278 101227-276 CA101227-278
Enterokinase
Enzymes
Enterokinase is a specific protease that cleaves after lysine at its cleavage site Asp-Asp-Asp-Asp-Lys
It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate. Enterokinase will not cleave at site followed by proline.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty