Would you like to log in again?
K. lactis GG799 Competent Cells, New England Biolabs
Supplier: New England Biolabs
C1001S
101227-510EA 463 USD InStock
101227-510
K. lactis GG799 Competent Cells, New England Biolabs
Competent Cells
Chemically competent Kluyveromyces lactis cells suitable for transformation with a linearized pKLAC-series expression vector
GG799 cells are characterized by very high cell density growth and efficient expression of foreign proteins. GG799 cells have no genetic modifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty