Would you like to log in again?
Amylose Resin High Flow, New England Biolabs
Supplier: New England Biolabs
E8022L E8022S E8022L E8022S
101227-548EA 388 USD InStock
101227-548 101227-546 CA101227-548 CA101227-546
Amylose Resin High Flow, New England Biolabs
Protein Purification Protein Purification Resins
Amylose Resin High Flow is a cross-linked affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP)
This rigid matrix can be used in automated chromatography systems. DNASU is a central repository for plasmid clones and collections that may also be helpful.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty