Would you like to log in again?
M13mp18 Single-Stranded DNA, New England Biolabs
Supplier: New England Biolabs
N4040S N4040S
101227-718EA 48 USD InStock
101227-718 CA101227-718
M13mp18 Single-Stranded DNA, New England Biolabs
Vectors, Plasmids and Libraries
The single-stranded viral DNA is isolated from M13mp18
M13mp18 is a M13 lac phage vector which contains single HindIII, SphI, SbfI, PstI, SalI (AccI/ HincII), XbaI, BamHI, SmaI (XmaI), KpnI (Acc65I), SacI and EcoRI sites within the gene encoding β-Galactosidase.This DNA preparation is useful as a standard and has been tested as a template in the dideoxy-nucleotide termination method of sequencing DNA. M13mp18 phage is propagated in E. coli ER2738. The phage is purified by polyethylene glycol precipitation. The single-stranded DNA is then extracted with phenol.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty