Would you like to log in again?
Native PNGase F
Supplier: New England Biolabs
P0704S P0704S P0704L P0704L
101227-726EA 225 USD InStock
101227-726 101227-724 CA101227-726 CA101227-724
Native PNGase F
Enzymes
Peptide-N-Glycosidase F, also known as PNGase F, is an amidase which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid and complex oligosaccharides from N-linked glycoproteins
A glycerol-free version of PNGaseF is also offered for HPLC methods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty