Would you like to log in again?
PNGase F
Supplier: New England Biolabs
P0705S P0705S P0705L P0705L
101227-728EA 896 USD InStock
CA101227-728 101227-728 CA101227-730 101227-730
PNGase F
Enzymes
Peptide-N-Glycosidase F, also known as PNGase F, is an amidase and supplied glycerol free for optimal performance in HPLC intensive methods
  • Glycerol-free
PNGase F cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid and complex oligosaccharides from N-linked glycoproteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty