Would you like to log in again?
beta-1-3 Galactosidase
Supplier: New England Biolabs
P0726S P0726S
101227-738EA 153 USD InStock
CA101227-738 101227-738
beta-1-3 Galactosidase
Enzymes
β1-3 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of β1-3 and, at a much lower rate, β1-6 linked D-galactopyranosyl residues from oligosaccharides
The approximate kinetic data show > 100-fold preference for β1-3 over β1-6 linkages, and > 500-fold preference from β1-3 over β1-4 linkages.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty