Would you like to log in again?
Alpha 1-6 Mannosidase
Supplier: New England Biolabs
P0727S P0727S
101227-742EA 153 USD InStock
CA101227-742 101227-742
Alpha 1-6 Mannosidase
Enzymes
α1-6 Mannosidase is a highly specific exoglycosidase that removes unbranched α1-6 linked D­mannopyranosyl residues from oligosaccharides
When used in conjunction with α1-2,3 Mannosidase, the α1-6 Mannosidase will cleave α1-6 Mannose residues from branched carbohydrate substrates.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty